1-Piperidin-1-ylacetone

Supplier: Matrix Scientific

035810-500MG
101840-112EA 290.77 USD
101840-112
1-Piperidin-1-ylacetone
1-Piperidin-1-ylacetone

Hazard: Irritant

Formula: C₈H₁₅NO
MDL Number: MFCD09864582
CAS Number: 6784-61-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR