2-Hydrazino-1H-1,3-benzimidazole

Supplier: Matrix Scientific

Synonyms: 2-Hydrazinyl-1H-benzo[d]imidazole

035802-500MG
101840-096EA 24.62 USD
101840-096
2-Hydrazino-1H-1,3-benzimidazole
2-Hydrazino-1H-1,3-benzimidazole

Hazard: Irritant

Formula: C₇H₈N₄
MW: 148.17 g/mol
MDL Number: MFCD00513603
CAS Number: 15108-18-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR