2-Ethyl-1,3-thiazole-4-carboxylic acid

Supplier: Matrix Scientific

035798-500MG
101840-088EA 484.62 USD
101840-088
2-Ethyl-1,3-thiazole-4-carboxylic acid
2-Ethyl-1,3-thiazole-4-carboxylic acid

Hazard: Irritant

Formula: C₆H₇NO₂S
MDL Number: MFCD09258842
CAS Number: 769124-05-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR