N-Methyl-N-[(1,3,5-trimethyl-1H-pyrazol-4-yl)methyl]amine

Supplier: Matrix Scientific

035792-500MG
101840-076EA 193.85 USD
101840-076
N-Methyl-N-[(1,3,5-trimethyl-1H-pyrazol-4-yl)methyl]amine
N-Methyl-N-[(1,3,5-trimethyl-1H-pyrazol-4-yl)methyl]amine

Hazard: Irritant

Formula: C₈H₁₅N₃
MW: 153.23 g/mol
MDL Number: MFCD02056512
CAS Number: 514816-08-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR