(1-Morpholin-4-ylcyclopentyl)methylamine

Supplier: Matrix Scientific

035790-500MG
101840-072EA 403.08 USD
101840-072
(1-Morpholin-4-ylcyclopentyl)methylamine
(1-Morpholin-4-ylcyclopentyl)methylamine

Hazard: Irritant

Formula: C₁₀H₂₀N₂O
MDL Number: MFCD08703639
CAS Number: 444666-61-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR