(2-Phenyl-2H-1,2,3-triazol-4-yl)methylamine

Supplier: Matrix Scientific

035783-500MG
101840-058EA 546.15 USD
101840-058
(2-Phenyl-2H-1,2,3-triazol-4-yl)methylamine
(2-Phenyl-2H-1,2,3-triazol-4-yl)methylamine

Hazard: Irritant

Formula: C₉H₁₀N₄
MDL Number: MFCD08669767
CAS Number: 912761-28-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR