(1,3,5-Trimethyl-1H-pyrazol-4-yl)methylamine

Supplier: Matrix Scientific

Synonyms: 4-(Aminomethyl)-1,3,5-trimethyl-1H-pyrazole

035781-500MG
101840-054EA 546.15 USD
101840-054
(1,3,5-Trimethyl-1H-pyrazol-4-yl)methylamine
(1,3,5-Trimethyl-1H-pyrazol-4-yl)methylamine

Hazard: Irritant

Formula: C₇H₁₃N₃
MW: 139.2 g/mol
MDL Number: MFCD02055811
CAS Number: 352018-93-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR