N-(2-(Trimethylsilyl)furo[3,2-b]pyridin-7-yl)pivalamide

Supplier: Matrix Scientific

035778-500MG 035778-100MG
101840-042EA 1090.77 USD
101840-042 101840-040
N-(2-(Trimethylsilyl)furo[3,2-b]pyridin-7-yl)pivalamide
N-(2-(Trimethylsilyl)furo[3,2-b]pyridin-7-yl)pivalamide

Hazard: Irritant

Formula: C₁₅H₂₂N₂O₂Si
MW: 290.44 g/mol
MDL Number: MFCD12026773
CAS Number: 1142192-62-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR