N-(2-Bromo-5-((hydroxyimino)methyl)pyridin-3-yl)pivalamide
Supplier: Matrix Scientific
035760-500MG
035760-100MG
101839-968EA
364.62
USD
101839-968
101839-970
N-(2-Bromo-5-((hydroxyimino)methyl)pyridin-3-yl)pivalamide
N-(2-Bromo-5-((hydroxyimino)methyl)pyridin-3-yl)pivalamide
Hazard: Irritant
Formula:
C₁₁H₁₄BrN₃O₂ MW: 300.16 g/mol |
MDL Number:
MFCD12026765 CAS Number: 1142192-46-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...