N-(2-Methoxypyridin-3-yl)pivalamide
Supplier: Matrix Scientific
035759-100MG
035759-500MG
101839-966EA
1090.77
USD
101839-966
101839-964
N-(2-Methoxypyridin-3-yl)pivalamide
N-(2-Methoxypyridin-3-yl)pivalamide
Hazard: Irritant
Formula:
C₁₁H₁₆N₂O₂ |
MDL Number:
MFCD12026764 CAS Number: 125867-19-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...