2-Bromo-5-iodopyridin-3-yl-tert-butyl carbonate

Supplier: Matrix Scientific

035752-100MG 035752-500MG
101839-936EA 364.62 USD
101839-936 101839-938
2-Bromo-5-iodopyridin-3-yl-tert-butyl carbonate
2-Bromo-5-iodopyridin-3-yl-tert-butyl carbonate

Hazard: Irritant

Formula: C₁₀H₁₁BrINO₃
MDL Number: MFCD12026757
CAS Number: 1142192-37-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR