N-(2-Bromo-5-hydroxypyridin-3-yl)pivalamide
Supplier: Matrix Scientific
035747-500MG
035747-100MG
101839-916EA
364.62
USD
101839-916
101839-918
N-(2-Bromo-5-hydroxypyridin-3-yl)pivalamide
N-(2-Bromo-5-hydroxypyridin-3-yl)pivalamide
Hazard: Irritant
Formula:
C₁₀H₁₃BrN₂O₂ |
MDL Number:
MFCD12026752 CAS Number: 1142192-30-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...