4-Amino-5-methoxynicotinonitrile

Supplier: Matrix Scientific

035737-100MG 035737-500MG
101839-878EA 1090.77 USD
101839-878 101839-876
4-Amino-5-methoxynicotinonitrile
4-Amino-5-methoxynicotinonitrile

Hazard: Irritant

Formula: C₇H₇N₃O
MDL Number: MFCD12026748
CAS Number: 1142192-15-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR