N-(2-Chloro-6-(dimethoxymethyl)pyridin-3-yl)pivalamide

Supplier: Matrix Scientific

035731-500MG 035731-100MG
101839-852EA 364.62 USD
101839-852 101839-854
N-(2-Chloro-6-(dimethoxymethyl)pyridin-3-yl)pivalamide
N-(2-Chloro-6-(dimethoxymethyl)pyridin-3-yl)pivalamide

Hazard: Irritant

Formula: C₁₃H₁₉ClN₂O₃
MW: 286.76 g/mol
MDL Number: MFCD11857732
CAS Number: 1142192-06-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR