tert-Butyl (6-chloro-5-pivalamidopyridin-2-yl)methylcarbamate

Supplier: Matrix Scientific

035728-100MG 035728-500MG
101839-840EA 364.62 USD
101839-840 101839-842
tert-Butyl (6-chloro-5-pivalamidopyridin-2-yl)methylcarbamate
tert-Butyl (6-chloro-5-pivalamidopyridin-2-yl)methylcarbamate

Hazard: Irritant

Formula: C₁₆H₂₄ClN₃O₃
MW: 341.84 g/mol
MDL Number: MFCD11857729
CAS Number: 1142192-00-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR