N-(6-((tert-Butyldimethylsilyloxy)methyl)-2-chloropyridin-3-yl)pivalamide

Supplier: Matrix Scientific

035722-500MG 035722-100MG
101839-816EA 364.62 USD
101839-816 101839-818
N-(6-((tert-Butyldimethylsilyloxy)methyl)-2-chloropyridin-3-yl)pivalamide
N-(6-((tert-Butyldimethylsilyloxy)methyl)-2-chloropyridin-3-yl)pivalamide

Hazard: Irritant

Formula: C₁₇H₂₉ClN₂O₂Si
MDL Number: MFCD11857723
CAS Number: 1142191-94-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR