N-(2-Chloro-6-((hydroxyimino)methyl)pyridin-3-yl)pivalamide
Supplier: Matrix Scientific
035716-500MG
035716-100MG
101839-792EA
364.62
USD
101839-792
101839-794
N-(2-Chloro-6-((hydroxyimino)methyl)pyridin-3-yl)pivalamide
N-(2-Chloro-6-((hydroxyimino)methyl)pyridin-3-yl)pivalamide
Hazard: Irritant
Formula:
C₁₁H₁₄ClN₃O₂ MW: 255.71 g/mol |
MDL Number:
MFCD11857718 CAS Number: 1142191-88-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...