tert-Butyl (4-cyano-5-methoxypyridin-3-yl)methylcarbamate

Supplier: Matrix Scientific

035714-100MG 035714-500MG
101839-784EA 364.62 USD
101839-784 101839-786
tert-Butyl (4-cyano-5-methoxypyridin-3-yl)methylcarbamate
tert-Butyl (4-cyano-5-methoxypyridin-3-yl)methylcarbamate

Hazard: Irritant

Formula: C₁₃H₁₇N₃O₃
MW: 263.3 g/mol
MDL Number: MFCD11857716
CAS Number: 1142191-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR