N-(6-Allyl-2-chloropyridin-3-yl)pivalamide

Supplier: Matrix Scientific

035708-500MG 035708-100MG
101839-760EA 364.62 USD
101839-760 101839-762
N-(6-Allyl-2-chloropyridin-3-yl)pivalamide
N-(6-Allyl-2-chloropyridin-3-yl)pivalamide

Hazard: Irritant

Formula: C₁₃H₁₇ClN₂O
MDL Number: MFCD11857710
CAS Number: 1142191-77-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR