2,5-Diiodopyridin-3-ol

Supplier: Matrix Scientific

035701-100MG 035701-500MG
101839-732EA 364.62 USD
101839-732 101839-734
2,5-Diiodopyridin-3-ol
2,5-Diiodopyridin-3-ol

Hazard: Irritant

Formula: C₅H₃I₂NO
MDL Number: MFCD11857702
CAS Number: 1142191-68-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR