4-Formyl-5-methoxy-N-phenylnicotinamide

Supplier: Matrix Scientific

035693-500MG 035693-100MG
101839-702EA 1090.77 USD
101839-702 101839-700
4-Formyl-5-methoxy-N-phenylnicotinamide
4-Formyl-5-methoxy-N-phenylnicotinamide

Hazard: Irritant

Formula: C₁₄H₁₂N₂O₃
MDL Number: MFCD12026745
CAS Number: 1142191-55-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR