3-Dibenzylamino-2-fluoropropionic acid, benzylester ≥95%

Supplier: Matrix Scientific

035686-1G
101839-674EA 283.08 USD
101839-674
3-Dibenzylamino-2-fluoropropionic acid, benzylester ≥95%
3-Dibenzylamino-2-fluoropropionic acid, benzylester

Hazard: Irritant

Formula: C₂₄H₂₄FNO₂
MDL Number: MFCD07367686
CAS Number: 887352-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR