2-(1,1-Difluoroethyl)-3-methylpyrazine ≥95%

Supplier: Matrix Scientific

035670-1G
101839-618EA 241.54 USD
101839-618
2-(1,1-Difluoroethyl)-3-methylpyrazine ≥95%
2-(1,1-Difluoroethyl)-3-methylpyrazine

Hazard: Irritant

Formula: C₇H₈F₂N₂
MDL Number: MFCD12026742
CAS Number: 1171916-84-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR