5-Carboxy-2-chlorophenylboronic acid
Supplier: Matrix Scientific
Synonyms:
2-Chloro-5-carboxyphenylboronic acid
035636-5G
035636-1G
101839-580EA
32.31
USD
101839-580
101839-582
5-Carboxy-2-chlorophenylboronic acid
5-Carboxy-2-chlorophenylboronic acid
Hazard: Irritant/Refrigerate
Formula:
C₇H₆BClO₄ MW: 200.3800 g/mol |
MDL Number:
MFCD08689491 CAS Number: 913835-75-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...