3-(Methylthio)phenylboronic acid pinacolate

Supplier: Matrix Scientific

Synonyms: 3-Methylthiophenylboronic acid pinacol ester4,4,5,5-Tetramethyl-2-(3-(methylthio)phenyl)-1,3,2-dioxaborolane

035478-1G
101839-458EA 235.38 USD
101839-458
3-(Methylthio)phenylboronic acid pinacolate
3-(Methylthio)phenylboronic acid pinacolate

Hazard: Irritant

Formula: C₁₃H₁₉BO₂S
MW: 250.17 g/mol
MDL Number: MFCD05155220
CAS Number: 710348-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR