2-(Hydroxymethyl)phenylboronic acid

Supplier: Matrix Scientific

Synonyms: 2-(Hydroxymethyl)benzeneboronic acid(2-(Hydroxymethyl)phenyl)boronic acid

035466-1G
101839-442EA 20.7 USD
101839-442
2-(Hydroxymethyl)phenylboronic acid
2-(Hydroxymethyl)phenylboronic acid

Hazard: Irritant/Store in Freezer

Formula: C₇H₉BO₃
MW: 151.96 g/mol
MDL Number: MFCD02258966
CAS Number: 87199-14-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR