2-Fluoro-4-(trifluoromethyl)phenylboronic acid

Supplier: Matrix Scientific

Synonyms: 2-Fluoro-4-(trifluoromethyl)benzeneboronic acid

035445-25G
102663-672EA 246.15 USD
102663-672
2-Fluoro-4-(trifluoromethyl)phenylboronic acid
2-Fluoro-4-(trifluoromethyl)phenylboronic acid

Hazard: Irritant/Store in Freezer

Formula: C₇H₅BF₄O₂
MW: 207.92 g/mol
MDL Number: MFCD03094995
CAS Number: 503309-11-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR