2-Methyl-4-phenylthiazol ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Methyl-4-phenylthiazole

035276-1G
101839-334EA 83.08 USD
101839-334
2-Methyl-4-phenylthiazol ≥97%
2-Methyl-4-phenylthiazol

Hazard: Irritant

Formula: C₁₀H₉NS
MW: 175.25
MDL Number: MFCD00489998
CAS Number: 1826-16-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR