3-Cyclopentylacrylonitrile

035272-250MG
101839-324EA 676.71 USD
101839-324
3-Cyclopentylacrylonitrile
3-Cyclopentylacrylonitrile

Hazard: Irritant

Formula: C₈H₁₁N
MDL Number: MFCD11857755
CAS Number: 591769-05-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR