6-Bromo-2-chloro-4-iodopyridin-3-amine

Supplier: Matrix Scientific

035263-250MG
101839-306EA 606.15 USD
101839-306
6-Bromo-2-chloro-4-iodopyridin-3-amine
6-Bromo-2-chloro-4-iodopyridin-3-amine

Hazard: Irritant

Formula: C₅H₃BrClIN₂
MDL Number: MFCD11857701
CAS Number: 1138444-29-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR