Methyl-3-(5,6-dimethoxypyridin-2-yl)acrylate

Supplier: Matrix Scientific

035260-250MG
101839-300EA 604.62 USD
101839-300
Methyl-3-(5,6-dimethoxypyridin-2-yl)acrylate
Methyl-3-(5,6-dimethoxypyridin-2-yl)acrylate

Hazard: Irritant

Formula: C₁₁H₁₃NO₄
MDL Number: MFCD11857698
CAS Number: 1138444-25-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR