3-Methyl-3H-imidazo[4,5-b]pyridine-6-carboxylic acid

Supplier: Matrix Scientific

035259-250MG
101839-298EA 109.23 USD
101839-298
3-Methyl-3H-imidazo[4,5-b]pyridine-6-carboxylic acid
3-Methyl-3H-imidazo[4,5-b]pyridine-6-carboxylic acid

Hazard: Irritant

Formula: C₈H₇N₃O₂
MDL Number: MFCD11857697
CAS Number: 1138444-24-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR