2-(Chloromethyl)-1-propyl-1H-imidazo[4,5-c]pyridine hydrochloride
Supplier: Matrix Scientific
035258-250MG
101839-296EA
606.15
USD
101839-296
2-(Chloromethyl)-1-propyl-1H-imidazo[4,5-c]pyridine hydrochloride
2-(Chloromethyl)-1-propyl-1H-imidazo[4,5-c]pyridine hydrochloride
Hazard: Irritant
Formula:
C₁₀H₁₃Cl₂N₃ |
MDL Number:
MFCD11857695 CAS Number: 1092381-05-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...