5-Methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)nicotinonitrile

Supplier: Matrix Scientific

035250-250MG
101839-280EA 489.23 USD
101839-280
5-Methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)nicotinonitrile
5-Methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)nicotinonitrile

Hazard: Irritant

Formula: C₁₃H₁₇BN₂O₃
MDL Number: MFCD11857687
CAS Number: 1247726-68-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR