6-Bromofuro[3,2-b]pyridine

Supplier: Matrix Scientific

035249-250MG
101839-278EA 609.23 USD
101839-278
6-Bromofuro[3,2-b]pyridine
6-Bromofuro[3,2-b]pyridine

Hazard: Irritant

Formula: C₇H₄BrNO
MDL Number: MFCD11857686
CAS Number: 934330-61-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR