2,6-Diiodo-3-methoxypyridine

Supplier: Matrix Scientific

035246-250MG
101839-272EA 604.62 USD
101839-272
2,6-Diiodo-3-methoxypyridine
2,6-Diiodo-3-methoxypyridine

Hazard: Irritant

Formula: C₆H₅I₂NO
MDL Number: MFCD11857683
CAS Number: 437709-98-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR