4-Iodo-5-methoxynicotinonitrile

Supplier: Matrix Scientific

035239-250MG
101839-256EA 606.15 USD
101839-256
4-Iodo-5-methoxynicotinonitrile
4-Iodo-5-methoxynicotinonitrile

Hazard: Irritant

Formula: C₇H₅IN₂O
MDL Number: MFCD11857678
CAS Number: 1138444-07-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR