6-Bromo-3-methyl-3H-imidazo[4,5-b]pyridine

Supplier: Matrix Scientific

035234-250MG
101839-246EA 78.46 USD
101839-246
6-Bromo-3-methyl-3H-imidazo[4,5-b]pyridine
6-Bromo-3-methyl-3H-imidazo[4,5-b]pyridine

Hazard: Irritant

Formula: C₇H₆BrN₃
MDL Number: MFCD08692133
CAS Number: 37805-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR