Methyl-3-(5-bromo-3-methoxypyridin-2-yl)acrylate

Supplier: Matrix Scientific

035228-250MG
101839-234EA 606.15 USD
101839-234
Methyl-3-(5-bromo-3-methoxypyridin-2-yl)acrylate
Methyl-3-(5-bromo-3-methoxypyridin-2-yl)acrylate

Hazard: Irritant

Formula: C₁₀H₁₀BrNO₃
MDL Number: MFCD11857668
CAS Number: 1138443-99-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR