Methyl-3-(5-methoxypyridin-3-yl)acrylate

Supplier: Matrix Scientific

035227-250MG
101839-232EA 609.23 USD
101839-232
Methyl-3-(5-methoxypyridin-3-yl)acrylate
Methyl-3-(5-methoxypyridin-3-yl)acrylate

Hazard: Irritant

Formula: C₁₀H₁₁NO₃
MDL Number: MFCD11857667
CAS Number: 1000896-01-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR