5-(Dimethoxymethyl)-2,3-dimethoxypyridine

Supplier: Matrix Scientific

035226-250MG
101839-230EA 610.77 USD
101839-230
5-(Dimethoxymethyl)-2,3-dimethoxypyridine
5-(Dimethoxymethyl)-2,3-dimethoxypyridine

Hazard: Irritant

Formula: C₁₀H₁₅NO₄
MDL Number: MFCD11857665
CAS Number: 1138443-97-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR