2,3-Dimethoxy-6-methylpyridine

Supplier: Matrix Scientific

035224-250MG
101839-226EA 604.62 USD
101839-226
2,3-Dimethoxy-6-methylpyridine
2,3-Dimethoxy-6-methylpyridine

Hazard: Irritant

Formula: C₈H₁₁NO₂
MDL Number: MFCD11857663
CAS Number: 861019-58-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR