5,6-Dimethoxypyridine-2-carbonitrile

Supplier: Matrix Scientific

Synonyms: 5,6-Dimethoxypicolinonitrile

035222-250MG
101839-222EA 318.46 USD
101839-222
5,6-Dimethoxypyridine-2-carbonitrile
5,6-Dimethoxypyridine-2-carbonitrile

Hazard: Irritant

Formula: C₈H₈N₂O₂
MW: 164.1613 g/mol
MDL Number: MFCD11857661
CAS Number: 1112851-50-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR