5,6-Dimethoxynicotinaldehyde oxime

Supplier: Matrix Scientific

035220-250MG
101839-218EA 604.62 USD
101839-218
5,6-Dimethoxynicotinaldehyde oxime
5,6-Dimethoxynicotinaldehyde oxime

Hazard: Irritant

Formula: C₈H₁₀N₂O₃
MDL Number: MFCD11857659
CAS Number: 1138443-93-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR