3-(Dimethoxymethyl)-5-methoxy-4-(trimethylsilyl)pyridine

Supplier: Matrix Scientific

035219-250MG
101839-216EA 606.15 USD
101839-216
3-(Dimethoxymethyl)-5-methoxy-4-(trimethylsilyl)pyridine
3-(Dimethoxymethyl)-5-methoxy-4-(trimethylsilyl)pyridine

Hazard: Irritant

Formula: C₁₂H₂₁NO₃Si
MDL Number: MFCD11857658
CAS Number: 1138443-91-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR