(5,6-Dimethoxypyridin-3-yl)methanol

Supplier: Matrix Scientific

035218-250MG
101839-214EA 604.62 USD
101839-214
(5,6-Dimethoxypyridin-3-yl)methanol
(5,6-Dimethoxypyridin-3-yl)methanol

Hazard: Irritant

Formula: C₈H₁₁NO₃
MDL Number: MFCD11857657
CAS Number: 1138443-89-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR