5-Bromo-2-(dimethoxymethyl)-3-methoxypyridine

Supplier: Matrix Scientific

035215-250MG
101839-208EA 606.15 USD
101839-208
5-Bromo-2-(dimethoxymethyl)-3-methoxypyridine
5-Bromo-2-(dimethoxymethyl)-3-methoxypyridine

Hazard: Irritant

Formula: C₉H₁₂BrNO₃
MDL Number: MFCD11857654
CAS Number: 1138443-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR