5-Bromo-2-((tert-butyldimethylsilyloxy)methyl)-3-methoxypyridine

Supplier: Matrix Scientific

035214-250MG
101839-206EA 607.69 USD
101839-206
5-Bromo-2-((tert-butyldimethylsilyloxy)methyl)-3-methoxypyridine
5-Bromo-2-((tert-butyldimethylsilyloxy)methyl)-3-methoxypyridine

Hazard: Irritant

Formula: C₁₃H₂₂BrNO₂Si
MDL Number: MFCD11857653
CAS Number: 1138443-85-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR