2,3-Dimethoxy-5-(trimethylsilyl)pyridine

Supplier: Matrix Scientific

035213-250MG
101839-204EA 610.77 USD
101839-204
2,3-Dimethoxy-5-(trimethylsilyl)pyridine
2,3-Dimethoxy-5-(trimethylsilyl)pyridine

Hazard: Irritant

Formula: C₁₀H₁₇NO₂Si
MDL Number: MFCD11857652
CAS Number: 1131335-75-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR