(5-Bromo-3-methoxypyridin-2-yl)methanol

Supplier: Matrix Scientific

035207-250MG
101839-192EA 606.15 USD
101839-192
(5-Bromo-3-methoxypyridin-2-yl)methanol
(5-Bromo-3-methoxypyridin-2-yl)methanol

Hazard: Irritant

Formula: C₇H₈BrNO₂
MDL Number: MFCD11857646
CAS Number: 1087659-32-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR